Product Name
Tetrahydro-4H-thiopyran-4-one 99%
Product Number
155160
Product Brand
ALDRICH
CAS Number
1072-72-6
Molecular Weight
116.18
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Crystals
PURITY (GC AREA %)≥ 98.5 %
CARBON CONTENT50.9 - 52.5 %
SULFUR CONTENT (S/C-ANALYSIS)27.2 - 28.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD72 MONTHS