Product Name
Dimethyl 2-oxoheptylphosphonate technical grade, 85%
Product Number
157937
Product Brand
ALDRICH
CAS Number
36969-89-8
Molecular Weight
222.22
Test
Specification
APPEARANCECOLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID83.0% (MINIMUM)
COLOR TEST20 APHA (MAXIMUM)
 APPROVED AUGUST 7, 2008 RJM