Product Name
2-Bromoanisole 97%
Product Number
159239
Product Brand
ALDRICH
CAS Number
578-57-4
Molecular Weight
187.03
Test
Specification
APPEARANCE (COLOR)Colorless to Faint Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE