Product Name
4-Tritylaniline 97%
Product Number
159506
Product Brand
ALDRICH
CAS Number
22948-06-7
Molecular Weight
335.44
Test
Specification
APPEARANCE (COLOR)White to Beige and Faint Grey to Grey
APPEARANCE (FORM)Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
TITRATION (NT) HCLO4 0.1M96.5 - 103.5 %
PURITY (HPLC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE