Product Name
Phenylmethanesulfonyl chloride 98%
Product Number
159719
Product Brand
ALDRICH
CAS Number
1939-99-7
Molecular Weight
190.65
Test
Specification
APPEARANCE (COLOR)White to Light Yellow and Faint Beige to Light Beige and Faint Brown to Light Brown
APPEARANCE (FORM)Powder or Crystals or Granules
TITRATION97.5 - 102.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD72 MONTHS