Product Name
1,12-Dibromododecane 98%
Product Number
174866
Product Brand
ALDRICH
CAS Number
3344-70-5
Molecular Weight
328.13
Test
Specification
APPEARANCE (COLOR)White to Beige
APPEARANCE (FORM)Powder or Solid
TITRATION97.5 - 102.5 %
TITRATION (METHOD)BROMINE COMBUSTION
PURITY (GC AREA %)≥ 97.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE