Product Name
4-Cyanophenyl 4-butylbenzoate ≥99%
Product Number
180327
Product Brand
ALDRICH
CAS Number
38690-77-6
Molecular Weight
279.33
Test
Specification
APPEARANCEWHITE POWDER OR CRYSTALS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 76.7%-78.2%
 NITROGEN 4.8%- 5.2%
GAS LIQUID99.0% (MINIMUM)
 APPROVED AUGUST 5, 2008 RJM