Product Name
Cumene 99%
Product Number
185795
Product Brand
ALDRICH
CAS Number
98-82-8
Molecular Weight
120.19
Test
Specification
APPEARANCECOLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE AND STANDARD AS
 ILLUSTRATED ON PAGE 932A OF EDITION I,
 VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR
 SPECTRA".
GAS LIQUID98.5% (MINIMUM)
 APPROVED DECEMBER 20, 1993 JSB