Product Name
3,4-Dimethoxyphenol 97%
Product Number
194468
Product Brand
ALDRICH
CAS Number
2033-89-8
Molecular Weight
154.16
Test
Specification
Appearance (Color)Conforms to Requirements
 Off White to Brown
Appearance (Form)Powder
Infrared spectrumConforms to Structure
Purity (GC)≥96.5 %