Product Name
6-Aminopyridine-3-carboxylic acid 97%
Product Number
216879
Product Brand
ALDRICH
CAS Number
3167-49-5
Molecular Weight
138.12
Test
Specification
APPEARANCEWHITE TO TAN POWDER, CRYSTALS OR CRYSTALLINE
 POWDER AND/OR CHUNKS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION96.5% - 103.5% (WITH HCLO4)
HIGH PRESSURE LIQUID96.5% (MINIMUM)
 APPROVED SEPTEMBER 24, 2007 BTR