Product Name
Aminophyllin hydrate 99%
Product Number
216895
Product Brand
ALDRICH
CAS Number
317-34-0
Molecular Weight
420.43 (anhydrous basis)
Test
Specification
 ILLUSTRATED ON PAGE 709D OF EDITION I,
 VOLUME 2 OF "THE ALDRICH LIBRARY OF FT-IR
 SPECTRA".
ELEMENTAL ANALYSISCARBON 45.0% (MINIMUM) (DRIED BASIS)
 NITROGEN 32.8% (MINIMUM) (DRIED BASIS)
TITRATIONTYPICALLY CA. 5% H2O
THIN-LAYERCONSISTENT WITH 99% PURITY
SOLUBILITY25 MG/ML H2O; CLEAR, COLORLESS SOLUTION
 REVISED SEPTEMBER 28, 1998 JSB