Product Name
Ethyl methanesulfonate 99%
Product Number
220507
Product Brand
ALDRICH
CAS Number
62-50-0
Molecular Weight
124.16
Test
Specification
APPEARANCECOLORLESS TO YELLOW LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE AND STANDARD AS
 ILLUSTRATED ON PAGE 898B OF EDITION I,
 VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR
 SPECTRA".
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE AND STANDARD AS
 ILLUSTRATED ON PAGE 800 OF EDITION II OF
 "THE ALDRICH LIBRARY OF NMR SPECTRA".
 APPROVED SEPTEMBER 7, 1993 JSB