Product Name
(−)-α-Cedrene ≥95.0% (sum of enantiomers, GC)
Product Number
22133
Product Brand
ALDRICH
CAS Number
469-61-4
Molecular Weight
204.35
Test
Specification
APPEARANCE (COLOR)Colorless to Light Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 95.0 %
SPECIFIC ROTATION (20/D)-90.0 - -80.0 Degrees
CONCENTRATIONC=10 IN ETHANOL
REFRACTIVE INDEX N20/D1.497 - 1.499
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE