Product Name
N-Methylbenzamide ≥99%
Product Number
222798
Product Brand
ALDRICH
CAS Number
613-93-4
Molecular Weight
135.16
Test
Specification
Appearance (Color)White to Off-White
Appearance (Form)Conforms to Requirements
 Powder, Crystals or Chips
Infrared spectrumConforms to Structure
Carbon70.4 - 71.8 %
Nitrogen10.2 - 10.5 %
Purity (GC)≥99.0 %