Product Name
1,3,5-Tri-tert-butylbenzene 97%
Product Number
223778
Product Brand
ALDRICH
CAS Number
1460-02-2
Molecular Weight
246.43
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals or Fibers
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE