Product Name
(1S)-(−)-Camphanic chloride 98%
Product Number
226173
Product Brand
ALDRICH
CAS Number
39637-74-6
Molecular Weight
216.66
Test
Specification
APPEARANCEWHITE TO TAN TO GREY POWDER, CRYSTALS OR
 CRYSTALLINE POWDER
INFRARED SPECTRUMCONFORMS TO STRUCTURE
 OR
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
TITRATION97.5%-102.5% (WITH AGNO3 AFTER OXYGEN
 COMBUSTION)
OPTICAL ROTATION-18 +/- 1 DEGREES (C=2%, CCL4)
ENANTIOMERIC EXCESS98.5% (MINIMUM)
GAS LIQUID97.5% (MINIMUM)
 APPROVED APRIL 8, 2005 RJM