Product Name
N-Hydroxymaleimide 97%
Product Number
226351
Product Brand
ALDRICH
CAS Number
4814-74-8
Molecular Weight
113.07
Test
Specification
APPEARANCE (COLOR)White to Light Yellow and Faint Beige to Light Beige and Faint Brown to Light Brown
APPEARANCE (FORM)Powder or Crystals or Chunk(s)
PURITY (GC AREA %)≥ 96.5 %
CARBON CONTENT41.0 - 44.0 %
NITROGEN CONTENT11.9 - 12.8 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE