Product Name
Methoxyamine hydrochloride 98%
Product Number
226904
Product Brand
ALDRICH
CAS Number
593-56-6
Molecular Weight
83.52
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
ARGENTOMETR. TITRATION97.5 - 102.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE