Product Name
3-Methyl-2-oxazolidinone 99.5%
Product Number
228001
Product Brand
ALDRICH
CAS Number
19836-78-3
Molecular Weight
101.10
Test
Specification
APPEARANCE (COLOR)Colorless to Light Yellow
APPEARANCE (FORM)Liquid
COLOR (IN APHA)≤ 300 APHA
PURITY (GC AREA %)≥ 99.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE