Product Name
1-Hexanethiol 95%
Product Number
234192
Product Brand
ALDRICH
CAS Number
111-31-9
Molecular Weight
118.24
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 94.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE