Product Name
tert-Butyl nitrite 90%
Product Number
235385
Product Brand
ALDRICH
CAS Number
540-80-7
Molecular Weight
103.12
Test
Specification
Appearance (Color)Yellow
Appearance (Form)Liquid
Infrared spectrumConforms to Structure
Purity (GC)≥88,5 %