Product Name
L-Pipecolic acid Proline homolog., 99%
Product Number
237752
Product Brand
ALDRICH
CAS Number
3105-95-1
Molecular Weight
129.16
Test
Specification
APPEARANCEWHITE POWDER AND/OR LUMPS
INFRARED SPECTRUMCONFORMS TO STRUCTURE AND STANDARD AS
 ILLUSTRATED ON PAGE 585D OF EDITION I,
 VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR
 SPECTRA".
TITRATION98.5% - 101.5% (WITH HCLO4)
OPTICAL ROTATION-26(+/- 4) DEGREES (C=1%, H2O)
THIN-LAYERCONSISTENT WITH 99% PURITY
SOLUBILITY5% IN H2O; CLEAR TO SLIGHTLY HAZY, COLORLESS
 TO FAINT YELLOW SOLUTION
 APPROVED MAY 21, 1997 JSB