Product Name
Hydroquinone ≥99%
Product Number
240125
Product Brand
ALDRICH
CAS Number
123-31-9
Molecular Weight
110.11
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER, CRYSTALS,
 CRYSTALLINE POWDER, NEEDLES, FIBROUS POWDER,
 AND/OR SOLID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID99.0% (MINIMUM)
 REVISED FEBRUARY 8, 2005 RJM