Product Name
(Bromomethyl)cyclopropane 97%
Product Number
242403
Product Brand
ALDRICH
CAS Number
7051-34-5
Molecular Weight
135.00
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE