Product Name
Tetrabutylammonium (meta)periodate
Product Number
247057
Product Brand
ALDRICH
CAS Number
65201-77-6
Molecular Weight
433.37
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow
APPEARANCE (FORM)Powder or Crystals
TITRATIONTypically 29.28% I
TITRATION (METHOD)-
INFRARED SPECTRUMCONFORMS TO STRUCTURE