Product Name
Dimethyl N-cyanodithioiminocarbonate 90%
Product Number
249637
Product Brand
ALDRICH
CAS Number
10191-60-3
Molecular Weight
146.23
Test
Specification
APPEARANCE (COLOR)OFF-WHITE TO YELLOW OR TAN
APPEARANCE (FORM)Powder or Crystal or Chunk(s)
CARBON CONTENT29.2 - 36.5 %
NITROGEN CONTENT17.1 - 21.3 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE