Product Name
2-(Benzyloxy)ethanol 98%
Product Number
252867
Product Brand
ALDRICH
CAS Number
622-08-2
Molecular Weight
152.19
Test
Specification
APPEARANCECOLORLESS TO GOLD LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE AND STANDARD AS
 ILLUSTRATED ON PAGE 1157B OF EDITION I,
 VOLUME 1 OF "THE ALDRICH LIBRARY OF FT-IR
 SPECTRA".
GAS LIQUID97.5% (MINIMUM)
 APPROVED DECEMBER 12, 1996 JSB