Product Name
1-Hexadecanol ReagentPlus®, 99%
Product Number
258741
Product Brand
ALDRICH
CAS Number
36653-82-4
Molecular Weight
242.44
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)powder, chips, chunks or solid
PURITY (GC AREA %)≥ 98.5 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear to Very Slightly Hazy
SOLUBILITY (METHOD)1G IN 10ML ETHANOL
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD60 MONTHS