Product Name
Hexetidine, mixture of stereoisomers
Product Number
259187
Product Brand
ALDRICH
CAS Number
141-94-6
Molecular Weight
339.60
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 90.0 %
REMARKS ON GCAS A MIXTURE OF STERIOISOMERS
INFRARED SPECTRUMCONFORMS TO STRUCTURE