Product Name
Methyl 2-methylbenzoate 99%
Product Number
259985
Product Brand
ALDRICH
CAS Number
89-71-4
Molecular Weight
150.17
Test
Specification
APPEARANCE (COLOR)Colorless to Faint Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE