Product Name
1,3-Di-tert-butylbenzene 97%
Product Number
272051
Product Brand
ALDRICH
CAS Number
1014-60-4
Molecular Weight
190.32
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE