Product Name
2,4,6-Triphenylaniline 97%
Product Number
272515
Product Brand
ALDRICH
CAS Number
6864-20-6
Molecular Weight
321.41
Test
Specification
APPEARANCE (COLOR)Off White to Light Yellow to Light Red
APPEARANCE (FORM)Powder
TITRATION (NT) HCLO4 0.1M96.5 - 103.5 %
PURITY (HPLC AREA %)≥ 96.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE