Product Name
2-Chlorobenzoxazole 99%
Product Number
274089
Product Brand
ALDRICH
CAS Number
615-18-9
Molecular Weight
153.57
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow
APPEARANCE (FORM)Liquid
ARGENTOMETR. TITRATION98.5 - 101.5 %
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE