Product Name
4-Nitrophenyl 3-bromo-2,2-diethoxypropionate 97%
Product Number
277274
Product Brand
ALDRICH
CAS Number
87224-03-1
Molecular Weight
362.17
Test
Specification
APPEARANCEWHITE TO LIGHT YELLOW CRYSTALLINE POWDER OR
 CRYSTALLINE SOLID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION96.5% - 103.5% (WITH AGNO3 AFTER OXYGEN
 COMBUSTION)
THIN-LAYERCONSISTENT WITH 97% PURITY
 APPROVED FEBRUARY 5, 2001 JSB