Product Name
Methyl heptafluorobutyrate 99%
Product Number
282294
Product Brand
ALDRICH
CAS Number
356-24-1
Molecular Weight
228.06
Test
Specification
APPEARANCECOLORLESS LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
GAS LIQUID98.5% (MINIMUM)
 APPROVED AUGUST 4, 2008 RJM