Product Name
1,8-Dimethylnaphthalene 95%
Product Number
282537
Product Brand
ALDRICH
CAS Number
569-41-5
Molecular Weight
156.22
Test
Specification
APPEARANCEOFF-WHITE TO TAN CRYSTALS
INFRARED SPECTRUMCONFORMS TO STRUCTURE AND STANDARD.
GAS LIQUID94.5% (MINIMUM)
 APPROVED JANUARY 5, 1994 JSB