Product Name
L-Lysine hydrate 97%
Product Number
282677
Product Brand
ALDRICH
CAS Number
199926-21-1
Molecular Weight
146.19 (anhydrous basis)
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow to Faint Brown
APPEARANCE (FORM)Powder or Powder with Chunk(s)
TITRATION (NT) HCLO4 0.1M96.5 - 103.5 %
OPTICAL ROTATION22.0 ± 3.0 Degrees
CONCENTRATIONC=2, 6N HCL
INFRARED SPECTRUMCONFORMS TO STRUCTURE