Product Name
Magnesium bis(monoperoxyphthalate) hexahydrate 80%, technical grade
Product Number
283207
Product Brand
ALDRICH
CAS Number
Molecular Weight
494.64
Test
Specification
Appearance (Form)Conforms to Requirements
 Powder or Crystals
Infrared spectrumConforms to Structure
Proton NMR spectrumConforms to Structure
Titration by Na2S2O3≥78.0 %