Product Name
5,10,15,20-Tetraphenyl-21H,23H-porphine vanadium(IV) oxide
Product Number
283649
Product Brand
ALDRICH
CAS Number
14705-63-6
Molecular Weight
679.66
Test
Specification
Appearance (Form)Conforms to Requirements
 Powder, Crystals, Crystalline Powder and/or Chunks
Infrared spectrumConforms to Structure
Wave Length420 - 426 nm
Extinction Coefficient≥350000
 in Dichloromethane
Carbon68.4 - 87.1 %
Nitrogen7.2 - 9.2 %
Purity (HPLC)≥88.0 %