Product Name
β-D-Glucose pentaacetate 98%
Product Number
285943
Product Brand
ALDRICH
CAS Number
604-69-3
Molecular Weight
390.34
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder
PURITY (GC AREA %)≥ 97.5 %
OPTICAL ROTATION4.2 ± 1.0 Degrees
CONCENTRATIONC=1%, CHCL3
SOLUBILITY (COLOR)Colorless to Faint Yellow
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)50MG/ML, CHCL3
CARBON CONTENT48.0 - 50.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE