Product Name
Phenylpyruvic acid 98%
Product Number
286958
Product Brand
ALDRICH
CAS Number
156-06-9
Molecular Weight
164.16
Test
Specification
APPEARANCE (COLOR)White to Light Yellow
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) NAOH 0.1M97.5 - 102.5 %
PURITY (HPLC AREA %)≥ 97.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE