Product Name
Hydroxyapatite reagent grade, powder, synthetic
Product Number
289396
Product Brand
ALDRICH
CAS Number
12167-74-7
Molecular Weight
502.31
Test
Specification
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms to Structure
Complexiometric EDTA: % Ca%