Product Name
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane 98%
Product Number
291110
Product Brand
ALDRICH
CAS Number
23978-09-8
Molecular Weight
376.49
Test
Specification
Appearance (Color)White to Off-White
Appearance (Form)Conforms to Requirement
 Crystals or Powder
Infrared spectrumConforms to Structure
Purity (Titration by HCL04)97.5 - 102.5 %