Product Name
4,5-Difluoro-2-nitroaniline 98%
Product Number
292478
Product Brand
ALDRICH
CAS Number
78056-39-0
Molecular Weight
174.10
Test
Specification
Appearance (Color)Conforms to Requirements
 Dark Yellow to Orange
Appearance (Form)Conforms to Requirements
 Fibers, Powder or Crystals
Infrared spectrumConforms to Structure
 Dark Yellow to Orange
Carbon40.4 - 42.4 %
Nitrogen15.7 - 16.5 %
Purity (GC)≥97.5 %