Product Name
2-(2-Thiazolylazo)-p-cresol 97%
Product Number
294543
Product Brand
ALDRICH
CAS Number
1823-44-5
Molecular Weight
219.26
Test
Specification
APPEARANCEYELLOW TO ORANGE TO BROWN POWDER OR
 CRYSTALLINE POWDER
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 52.8%-56.7%
 NITROGEN 18.5%-19.8%
THIN-LAYERCONSISTENT WITH 97% PURITY
 OR
HIGH PRESSURE LIQUID96.5% (MINIMUM)
SOLUBILITY50MG/ML, ETHANOL:CHCL3 (1:1); CLEAR, RED TO
 BROWN SOLUTION
 APPROVED OCTOBER 25, 2007 RJM