Product Name
Furfuryl methyl sulfide 97%
Product Number
303763
Product Brand
ALDRICH
CAS Number
1438-91-1
Molecular Weight
128.19
Test
Specification
APPEARANCELIGHT TO DARK BROWN LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE AND STANDARD AS
 ILLUSTRATED ON PAGE 1454B OF EDITION I,
 VOLUME 3 OF "THE ALDRICH LIBRARY OF FT-IR
 SPECTRA".
GAS LIQUID96.5% (MINIMUM)
 APPROVED DECEMBER 28, 1995 JSB