Product Name
3,4-Hexanedione 95%
Product Number
306932
Product Brand
ALDRICH
CAS Number
4437-51-8
Molecular Weight
114.14
Test
Specification
APPEARANCE (COLOR)Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 94.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE