Product Name
(±)-Epinephrine 95%
Product Number
316741
Product Brand
ALDRICH
CAS Number
329-65-7
Molecular Weight
183.20
Test
Specification
APPEARANCEBEIGE TO TAN OR LIGHT BROWN POWDER
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
ELEMENTAL ANALYSISCARBON 55.5% - 62.5%
 NITROGEN 7.2% - 8.1%
 OR
TITRATION94.0% - 106.0% (WITH HCLO4)
THIN-LAYERCONSISTENT WITH 95% PURITY
SOLUBILITY5% IN 1M HCL; CLEAR, YELLOW TO YELLOW-TAN
 SOLUTION
 APPROVED FEBRUARY 27, 1997 JSB