Product Name
Methyl 2-(methylthio)butyrate 98%
Product Number
319678
Product Brand
ALDRICH
CAS Number
51534-66-8
Molecular Weight
148.22
Test
Specification
APPEARANCECOLORLESS TO VERY FAINT YELLOW LIQUID
INFRARED SPECTRUMCONFORMS TO STRUCTURE AND STANDARD AS
 ILLUSTRATED ON PAGE 730D OF EDITION I,
 VOLUME 3 OF "THE ALDRICH LIBRARY OF FT-IR
 SPECTRA".
GAS LIQUID97.5% (MINIMUM)
 APPROVED SEPTEMBER 8, 1998 JSB