Product Name
2,5-Bis(trifluoromethyl)aniline 99%
Product Number
331546
Product Brand
ALDRICH
CAS Number
328-93-8
Molecular Weight
229.12
Test
Specification
APPEARANCE (COLOR)Colorless to Faint Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE